SY3371477238 Synthonix Corporation

2-Cyclopropylmethoxy-4-fluorophenylboronic acid pinacol ester

  • MDL number MFCD13152010
  • Cas Number : 1185836-96-7
  • Molecular Weight : 292.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/5461.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/5461.html
pictograms GHS05, GHS07
hstatements H302, H315, H318, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart