SY3371477245 Synthonix Corporation

4-Chloro-3-fluoropyridine hydrochloride

  • MDL number MFCD03092936
  • Cas Number : 119229-74-2
  • Molecular Weight : 168

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/5574.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/5574.html
pictograms GHS05, GHS07
hstatements H302, H315, H318, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart