SY3371477264 Synthonix Corporation

6-Chloro-3-iodoimidazo[1,2-a]pyridine

  • MDL number MFCD06739217
  • Cas Number : 885275-59-2
  • Molecular Weight : 278.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/5921.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/5921.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart