SY3371477313 Synthonix Corporation

[3-Chloro-2-(1-ethoxyethoxy)propyl] acetate

  • MDL number MFCD22421689
  • Cas Number : 85328-35-4
  • Molecular Weight : 224.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 94
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/7025.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/7025.html
pictograms GHS07
hstatements H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart