SY3371477323 Synthonix Corporation

1,3-Dihydro-2H-pyrrolo[3,2-c]pyridin-2-one

  • MDL number MFCD09832902
  • Cas Number : 134682-54-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/0077.html

Safety Information

pcodes P280 ,
MSDS https://synthonix.com/0077.html
hstatements H301 , H302 , H319 ,
Availability details for
Quantity
  • Availability
  • Add to Cart