SY3371477344 Synthonix Corporation

2,4-Dichloro-6-methyl-7H-pyrrolo[2,3-d]pyrimidine

  • MDL number MFCD13193622
  • Cas Number : 1192711-71-9
  • Molecular Weight : 202.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/0559.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/0559.html
pictograms GHS06, GHS07, GHS08, GHS09
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart