SY3371477357 Synthonix Corporation

3-(Dibenzylamino)oxetane-3-carbonitrile

  • MDL number MFCD18432775
  • Cas Number : 1021393-00-9
  • Molecular Weight : 278.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/0695.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/0695.html
pictograms GHS05, GHS07
hstatements H302, H315, H318, H335
Availability details for
Quantity
  • Availability
  • Add to Cart