SY3371477407 Synthonix Corporation

2-(1,1-Dimethylethyl)-1H-Imidazole

  • MDL number MFCD16618480
  • Cas Number : 36947-69-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/1577.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/1577.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart