SY3371477429 Synthonix Corporation

Diethyl (2-chlorothiazol-5-yl)methylphosphonate

  • MDL number MFCD06660117
  • Cas Number : 903130-74-5
  • Molecular Weight : 269.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/2062.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/2062.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart