SY3371477450 Synthonix Corporation

4,5-Dimethyl-2-(tributylstannyl)thiazole

  • MDL number MFCD09265485
  • Cas Number : 938181-92-1
  • Molecular Weight : 402.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 94
Storage temp. Referigrator
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/2605.html

Safety Information

signalword Danger
pcodes P261, P262, P264, P270, P273, P280
MSDS https://synthonix.com/2605.html
pictograms GHS06, GHS08, GHS09
hstatements H301, H312, H315, H319, H332, H410
Availability details for
Quantity
  • Availability
  • Add to Cart