SY3371477460 Synthonix Corporation

2-(N,N-Dimethylamino)-3-formyl-4-iodopyridine

  • MDL number MFCD09702016
  • Cas Number : 944709-71-1
  • Molecular Weight : 276.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/2699.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/2699.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart