SY3371477472 Synthonix Corporation

4(5)-Diiodo-1H-imidazole

  • MDL number MFCD00963878
  • Cas Number : 15813-09-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/2856.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/2856.html
hstatements H302 , H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart