SY3371477482 Synthonix Corporation

2,2-Dimethylethenylboronic acid

  • MDL number MFCD01074609
  • Cas Number : 14559-88-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/3007.html

Safety Information

pcodes P261 ,
MSDS https://synthonix.com/3007.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart