SY3371477506 Synthonix Corporation

2,6-Dichloro-7-methylpurine

  • MDL number MFCD00039690
  • Cas Number : 2273-93-0
  • Molecular Weight : 203.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/3699.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/3699.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart