SY3371477539 Synthonix Corporation

3-Ethynyl-1-tosylazetidin-3-ol

  • MDL number MFCD22421707
  • Cas Number : 1349199-60-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/7029.html

Safety Information

MSDS https://synthonix.com/7029.html
Availability details for
Quantity
  • Availability
  • Add to Cart