SY3371477589 Synthonix Corporation

4,4-Dimethoxytetrahydro-4H-pyran

  • MDL number MFCD00040530
  • Cas Number : 28218-71-5
  • Molecular Weight : 146.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/5824.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/5824.html
pictograms GHS07
hstatements H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart