SY3371477590 Synthonix Corporation

alpha,alpha-Dimethyl-2-thiazolemethanol

  • MDL number MFCD03211903
  • Cas Number : 16077-78-4
  • Molecular Weight : 143.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 94
Storage temp. Referigrator
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/5833.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/5833.html
pictograms GHS07
hstatements H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart