SY3371477616 Synthonix Corporation

1,9-Diazaspiro[5.5]undecane-1-carboxylic acid tert-butyl ester

  • MDL number MFCD12406520
  • Cas Number : 1158750-00-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/6013.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/6013.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart