SY3371477651 Synthonix Corporation

6,7-Dihydro-5(4H)-cyclopenta[1,3]dioxin-5-one

  • MDL number MFCD00075482
  • Cas Number : 102306-78-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/6292.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/6292.html
hstatements H302 , H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart