SY3371477657 Synthonix Corporation

1,3-Diphenylparabanic acid

  • MDL number MFCD00020868
  • Cas Number : 6488-59-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://synthonix.com/6405.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/6405.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart