SY3371477658 Synthonix Corporation

cis-4,4-Difluorooctahydrocyclopenta[c]pyrrole

  • MDL number MFCD12198728
  • Cas Number : 1251008-46-4
  • Molecular Weight : 147.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. Referigrator
Physical Form liquid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/6528.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/6528.html
pictograms GHS05, GHS07
hstatements H302, H315, H318, H335
Availability details for
Quantity
  • Availability
  • Add to Cart