SY3371477666 Synthonix Corporation

3,3-Dimethoxyoxetane

  • MDL number MFCD12545828
  • Cas Number : 922500-97-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/6643.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/6643.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart