SY3371477756 Synthonix Corporation

3-Ethylsulfonylphenylboronic acid

  • MDL number MFCD03095257
  • Cas Number : 845870-47-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin USA
Product Link https://synthonix.com/2423.html

Safety Information

pcodes P261 ,
MSDS https://synthonix.com/2423.html
hstatements H301 , H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart