SY3371477760 Synthonix Corporation

2-Ethoxy-5-(tributylstannyl)thiazole

  • MDL number MFCD09025788
  • Cas Number : 446285-61-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/2530.html

Safety Information

pcodes P273 , P280 ,
MSDS https://synthonix.com/2530.html
hstatements H301 , H312 , H315 , H319 ,
Availability details for
Quantity
  • Availability
  • Add to Cart