SY3371477767 Synthonix Corporation

Ethyl 4-Bromo-5-methoxy-1H-indole-2-carboxylate

  • MDL number MFCD09265488
  • Molecular Weight : 298.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/2655.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/2655.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart