SY3371477768 Synthonix Corporation

(Z)-1-Ethoxyethene-2-boronic acid pinacol ester

  • MDL number MFCD09702441
  • Cas Number : 219489-07-3
  • Molecular Weight : 198.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Freezer
Physical Form Liquid
Shipping Temperature Ice Pak
Country of Origin US
Product Link https://synthonix.com/2791.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/2791.html
pictograms GHS07
hstatements H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart