SY3371477778 Synthonix Corporation

4-Ethylthiophenylboronic acid

  • MDL number MFCD01318146
  • Cas Number : 145349-76-4
  • Molecular Weight : 182.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/3166.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/3166.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart