SY3371477779 Synthonix Corporation

Ethyl 2-amino-5-bromothiazole-4-carboxylate

  • MDL number MFCD06669909
  • Cas Number : 61830-21-5
  • Molecular Weight : 251.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/3187.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/3187.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart