SY3371477782 Synthonix Corporation

Ethyl 6-methylimidazo[2,1-b]thiazole-5-carboxylate

  • MDL number MFCD01060560
  • Cas Number : 57626-37-6
  • Molecular Weight : 210.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/3328.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/3328.html
pictograms GHS07
hstatements H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart