SY3371477784 Synthonix Corporation

Ethyl 2-chloro-4-ethoxypyridine-3-carboxylate

  • MDL number MFCD09702463
  • Cas Number : 1171502-57-0
  • Molecular Weight : 229.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/3746.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/3746.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart