SY3371477795 Synthonix Corporation

Ethyl 7-chlorothieno[3,2-b]pyridine-6-carboxylate

  • Cas Number : 83179-01-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin USA
Product Link https://synthonix.com/4282.html

Safety Information

MSDS https://synthonix.com/4282.html
Availability details for
Quantity
  • Availability
  • Add to Cart