SY3371477797 Synthonix Corporation

Ethyl 4-chloropyrrolo[1,2-f][1,2,4]triazine-6-carboxylate

  • MDL number MFCD08448163
  • Cas Number : 903129-94-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin USA
Product Link https://synthonix.com/4284.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/4284.html
hstatements H302 , H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart