SY3371477807 Synthonix Corporation

Ethyl 4-chloro-7H-pyrrolo[2,3-d]pyrimidine-6-carboxylate

  • Cas Number : 187725-00-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/5321.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/5321.html
hstatements H302 , H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart