SY3371477819 Synthonix Corporation

Ethyl 2-(2-(4-chlorophenyl)thiazol-4-yl)acetate

  • MDL number MFCD01765002
  • Cas Number : 20287-70-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/5519.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/5519.html
hstatements H302 , H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart