SY3371477821 Synthonix Corporation

2-Ethoxy-1-propene

  • MDL number MFCD00048576
  • Cas Number : 926-66-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/5573.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/5573.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart