SY3371477824 Synthonix Corporation

4-Ethenylpiperidine-1-carboxylic acid tert-butyl ester

  • MDL number MFCD11036128
  • Cas Number : 180307-56-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/6108.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/6108.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart