SY3371477857 Synthonix Corporation

3-Ethyl-3-oxetanamine hydrochloride

  • MDL number MFCD19688088
  • Cas Number : 1379288-48-8
  • Molecular Weight : 137.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/7159.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/7159.html
pictograms GHS07
hstatements H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart