SY3371477858 Synthonix Corporation

2-Ethynyl-5-methyl-pyridin-3-amine

  • MDL number MFCD22189990
  • Cas Number : 1394319-72-2
  • Molecular Weight : 132.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/8223.html

Safety Information

signalword Warning
pcodes P202, P261, P280
MSDS https://synthonix.com/8223.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart