SY3371477908 Synthonix Corporation

3-Formyl-7-methylindole

  • MDL number MFCD00047170
  • Cas Number : 4771-50-0
  • Molecular Weight : 159.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/2104.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/2104.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart