SY3371477928 Synthonix Corporation

2-Formyl-4-(tributylstannyl)thiazole

  • MDL number MFCD09025791
  • Cas Number : 231278-14-1
  • Molecular Weight : 402.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/2457.html

Safety Information

signalword Danger
pcodes P261, P262, P264, P270, P273, P280
MSDS https://synthonix.com/2457.html
pictograms GHS06, GHS07, GHS08, GHS09
hstatements H301, H302, H312, H315, H319, H332, H335, H410
Availability details for
Quantity
  • Availability
  • Add to Cart