SY3371477933 Synthonix Corporation

2-Fluoro-5-methyl-3-(tributylstannyl)pyridine

  • MDL number MFCD09025799
  • Cas Number : 1025745-87-2
  • Molecular Weight : 400.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/2538.html

Safety Information

signalword Danger
pcodes P261, P262, P264, P270, P273, P280
MSDS https://synthonix.com/2538.html
pictograms GHS05, GHS06, GHS07, GHS08
hstatements H301, H302, H312, H315, H318, H319, H332, H335, H410
Availability details for
Quantity
  • Availability
  • Add to Cart