SY3371477991 Synthonix Corporation

Fmoc-Tyr(tBu)-Bt

  • MDL number MFCD11850165
  • Cas Number : 1126433-42-8
  • Molecular Weight : 560.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/5412.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/5412.html
pictograms GHS07
hstatements H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart