SY3371478011 Synthonix Corporation

2-Fluorothioacetamide

  • MDL number MFCD19204804
  • Cas Number : 84350-43-6
  • Molecular Weight : 93.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-fluoroethanethioamide
Inchi Code 1S/C2H4FNS/c3-1-2(4)5/h1H2,(H2,4,5)
Inchi Key OHBURFVUAFMQBE-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Refrigerator
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/6787.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/6787.html
pictograms GHS05, GHS07
hstatements H302, H315, H318, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart