SY3371478049 Synthonix Corporation

4-Hydroxy-5-methylpyrrolo[1,2-f][1,2,4] triazine

  • MDL number MFCD12923948
  • Cas Number : 529508-54-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/1215.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/1215.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart