SY3371478071 Synthonix Corporation

5-Hydroxymethyl-4-methylimidazole hydrochloride

  • MDL number MFCD00040986
  • Cas Number : 38585-62-5
  • Molecular Weight : 148.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/2822.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/2822.html
pictograms GHS07
hstatements H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart