SY3371478097 Synthonix Corporation

4-Hydroxyisoquinoline

  • MDL number MFCD00234514
  • Cas Number : 3336-49-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/6069.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/6069.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart