SY3371478123 Synthonix Corporation

Imidazo[1,2-a]pyridine-7-carboxylic acid

  • MDL number MFCD09834781
  • Cas Number : 648423-85-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/0355.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/0355.html
hstatements H315 , H317 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart