SY3371478157 Synthonix Corporation

Indole-4-boronic acid

  • MDL number MFCD03095175
  • Cas Number : 220465-43-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/2051.html

Safety Information

pcodes P261 ,
MSDS https://synthonix.com/2051.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart