SY3371478170 Synthonix Corporation

3-Isopropoxy-4-(tributylstannyl)-1,2-cyclobutenedione

  • MDL number MFCD01319011
  • Cas Number : 129034-70-4
  • Molecular Weight : 429.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Freezer
Physical Form Liquid
Shipping Temperature Ice Pak
Country of Origin US
Product Link https://synthonix.com/2855.html

Safety Information

signalword Danger
pcodes P261, P262, P264, P270, P273, P280
MSDS https://synthonix.com/2855.html
pictograms GHS06, GHS07, GHS08, GHS09
hstatements H301, H312, H315, H319, H332, H335, H410
Availability details for
Quantity
  • Availability
  • Add to Cart