SY3371478174 Synthonix Corporation

Imidazo[1,2-a]pyridine-3-carboxaldehyde

  • MDL number MFCD07778354
  • Cas Number : 6188-43-8
  • Molecular Weight : 146.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Viscous
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/3368.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/3368.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart